Learn More
MilliporeSigma™ Supelco™ column Blank Kit for HPLC
Each column blank kit includes a precision-bore polished 316 stainless steel column blank, two zero dead volume external reducing unions (internal unions with 10 mm I.D. blanks) with nuts and ferrules for 1/16 in. O.D. tubing, and two 2 μm frits.
$242.60 - $362.16
Specifications
Material 316 L Stainless Steel
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
111010713
View Documents
MilliporeSigma™ Supelco™
59101
I.D. O.D. L 4.6 mm 1/4 in. 15 cm
Each for $242.60
Only null left
111016267
View Documents
MilliporeSigma™ Supelco™
59102U
I.D. O.D. L 4.6 mm 1/4 in. 25 cm
Each for $299.61
Only null left
111010261
View Documents
MilliporeSigma™ Supelco™
58217
I.D. O.D. L 10 mm 1/2 in. 25 cm
Each for $362.16
Only null left
Description
Description
Specifications
Specifications
316 L Stainless Steel
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'