CombiSolvent Fats Solvent, MilliporeSigma™
CombiSolvent Fats Solvent for volumetric Karl Fischer titration with one component reagents for fats Aquastar™.
Supplier:  Technidata 1880211002
Catalog No. M1880211002
Explore available promotions
Description
Description
Specifications
Specifications
9°C
0.83 g/cm3 (at 20°C)
Glass Bottle
1 L
5.5 (H2O)