missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One ComboPlate™ 24-well Protein Crystallization Plate for Hanging Drop Application

Ideal for hanging and sitting drop vapor diffusion within a standardized footprint. Greiner Bio-One ComboPlate™ 24-Well Protein Crystallization Plate for Hanging Drop Application is universal plate with lid for crystallography.

Supplier:  Greiner Bio-One 662150

Catalog No. 07000673


Explore available promotions

Description

Description

  • Virgin, high-clarity polystyrene combined with distortion-free bottom provides optical properties for microscopic inspection of crystal growth
  • Flattened raised ring around each well reduces risk of cross-contamination and makes it poss
Specifications

Specifications

Standard
4 x 6
Polystyrene
Non-sterile
Hanging or sitting drop crystallization
24/Cs.
Well: 16.3 mm
With Lid
Clear
24
Flat Bottom (F-well)
24
18.8 mm
Videos
SDS
Documents

Documents

Promotions

Promotions