Epredia™ Complete Manual Staining Assemblies 117 and 130
Accommodate up to 60 slides with Epredia™ Complete Manual Staining Assemblies 117 and 130, available with a non-autoclavable glass or stainless-steel dish.
$193.45 - $193.45
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
12-0
|
Epredia™
SH120 |
Rack Handle only for Assemblies 117 and 130 |
Each for $193.45
|
|
|||||
Description
Complete Manual Staining Assembly 117:
- Staining Dish 119: Stainless steel with inside dimensions of 7.95 x 3.35 x 2.95 in. (202 x 85 x 75mm); holds up to 750mL of solution with fully loaded staining rack.
- Staining Rack 120: Stainless steel; holds up to 60 slides; maximum slide size is 3 x 2 in. (75 x 50mm).
- Staining Dish Cover 118: Stainless steel
Complete Manual Staining Assembly 130:
- Staining Dish 131: Nonautoclavable glass with inside dimensions of 7.95 x 4.4 x 2.8 in. (202 x 112 x 72mm); holds up to 750mL of solution with fully loaded staining rack
- Staining Rack 120: Stainless steel; holds up to 60 slides; maximum slide size is 3 x 2 in. (75 x 50mm)
- Staining Dish Cover 132: Stainless steel