Supertek™ Compound Bar DFS Item
Two metals of different heat coefficients, brass and iron are laminated together and mounted in a wooden handle. This device demonstrates the unequal expansion of different metals by bending when heated.
Supplier:  Supertek™ PH20060
Catalog No. S012332
Explore available promotions
Description
Description
  • Two metals of different heat coefficients, brass and iron are laminated together.
  • Mounted in a wooden handle.
  • Demonstrates the unequal expansion of different metals by bending when heated.
Specifications
Specifications
Heat Conductometer
6+
1 Lab Group