Learn More
Concanavalin A
A plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts | CAS: 11028-71-0
$130.63 - $259.49
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61221MC
|
Thermo Scientific Chemicals
J61221MC |
100 mg |
Each for $130.63
|
|
|||||
|
AAJ61221MD
|
Thermo Scientific Chemicals
J61221MD |
250 mg |
Each for $259.49
|
|
|||||
Description
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsConcanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
Solubility
Slightly soluble in water and phosfate buffered saline.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 11028-71-0 | |
| MFCD00071069 | |
| ConA |
Specifications
| Concanavalin A | |
| 11028-71-0 | |
| 100 mg | |
| 14,2495 | |
| Slightly soluble in water and phosfate buffered saline. |
| Lyophilized Powder | |
| White | |
| MFCD00071069 | |
| ConA | |
| Concanavalin A |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 234-258-2
RTECSNumber : GK6890000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only