missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 22365220
Specifications
Conductivity Standard | |
7647-14-5,7732-18-5 | |
20 x 20 mL | |
100.0,0.07 | |
Odorless | |
0°C | |
Completely miscible |
1.413mS/cm (1413 μmho/cm) at 25°C | |
Pack | |
98.0,0.07 | |
Colorless | |
6.7 to 7.3 | |
100°C | |
Liquid |