Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.
Supplier:  METTLER TOLEDO 51350092
 View more versions of this product
Catalog No. 01912590
Explore available promotions
Description
Description
  • For daily verification: to monitor health of electrode and ensure measurement accuracy
  • For initial calibration of nominal cell constant electrodes
  • Be sure to screw cap tightly immediately after use (sachets minimize the chance of error due to environmental interaction)
  • Conductivity standards subject to change values when exposed to environment; water evaporation raises conductivity values
Specifications
Specifications
Bottle
250 mL
Colorless
Conductivity Electrodes
1413 μS/cm
77447-40-7,7732-18-5
Conductivity Standard
Conductivity Standard
6