missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Confined Space Entry Permit

25 permits per pad. The Brady™ Confined Space Entry Permit are OSHA-compliant forms.

$13.15 - $60.32

Specifications

Width (English) Page 8.5 in.
Length (English) Page 11 in.
Color Black on White
Dimensions (L x W) 8.5 x 11 in.
Quantity 25/Pad
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-103-330
View Documents
Brady™
65937
2-ply Carbonless
Pack of 25 Each for $60.32
Only null left
 
19-103-346
View Documents
Brady™
65936
20# bond paper
Each for $23.24
Only null left
 
19-103-331
View Documents
Brady™
EP1
Prinzing
Each for $13.15
Only null left
 
Description

Description

  • OSHA-compliant
  • 25 permits per pad
Specifications

Specifications

8.5 in.
Black on White
25/Pad
11 in.
8.5 x 11 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'