missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 26 Monoclonal Antibody (CX-1E8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Fisher Scientific 335800
Description
33-5800 was used in the immunofluorescence and western blot analysis to successfully detect Connexin26 in mouse liver sections and homogenate, respectively.
Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.Specifications
| Connexin 26 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide, pH 7.4 | |
| P21994, Q00977 | |
| Gjb2 | |
| A 13 amino acid synthetic peptide derived from the C-terminus of the mouse Connexin 26 protein. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (Frozen), Western Blot | |
| CX-1E8 | |
| Unconjugated | |
| Gjb2 | |
| AI325222, Cnx26, connexin 26, connexin-26, CX26, CXN-26, DFNA3, DFNA3A, DFNB1, DFNB1A, Gap junction beta-2 protein, gap junction channel protein connexin 26, gap junction membrane channel protein beta 2, gap junction protein beta 2, gap junction protein, beta 2, gap junction protein, beta 2, 26kDa, GJB2, Gjb-2, HID, KID, mutant gap junction protein beta 2, NSRD1, PPK | |
| Mouse | |
| Protein A | |
| RUO | |
| 14619, 394266 | |
| -20°C | |
| Liquid |