Contec™ Amplitude™ Zeta™ Cleanroom Wipes
Prepare for acid spills. The Contec™ Amplitude™ Zeta™ Specialty Cleanroom Wipes are excellent for use with acid spills or for use as a tray liner. Wipes are lot-numbered for traceability.
Supplier:  Contec™ AMZE0001
 View more versions of this product
Catalog No. 19-130-5923
Explore available promotions
Description
Description
Prepare for acid spills. The Contec™ Amplitude™ Zeta™ Specialty Cleanroom Wipes are excellent for use with acid spills or for use as a tray liner. Wipes are lot-numbered for traceability.
  • Cellulose/polypropylene nonwoven wipes
  • Thermally-laminated, quilted wipes feature polypropylene outer layers and a highly sorptive cellulose center
  • Quilted pattern allows for segregation by appearance
  • Excellent for acid spills 
  • Ideal for tray liners
  • Lot-numbered for traceability
  • Inner core of cellulose thermally sealed between two layers of meltblown polypropylene is sorptive
  • Strong resistance to acids
  • Three-ply quilted pattern allows for segregation by appearance
Specifications
Specifications
Specialty Cleanroom Wipe
20.3 cm
23 cm
ISO Class 6-8
Cleanroom
8 in.
9 in.
Polypropylene
8 x 9 in. (20.3 x 23 cm)