Contec™ Trolley and Double Bucket Systems
Features a stainless steel rolling cart with removable handle for easier autoclaving and more convenient storage. Contec™ Trolley and Double Bucket System offers multiple wringer choices to accommodate all Contec™ mop systems.
Supplier: Contec™ 2650
Description
Features a stainless steel rolling cart with removable handle for easier autoclaving and more convenient storage. Contec™ Trolley and Double Bucket System offers multiple wringer choices to accommodate all Contec™ mop systems.
- Durable, lightweight and compact design improves maneuverability and reduces storage space
- Four casters; two fixed and two swivel and lock easily with the touch of a foot
- System can be used with two or three buckets, depending upon the required cleaning protocols; extendable frame accommodates additional bucket
- Included stainless steel connecting kit prevents drips between buckets
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- For maximum flexibility, wringers are ordered separately
Specifications
Polypropylene, Stainless Steel | |
36 in. | |
56 cm |
Autoclavable | |
91 cm | |
2 x 6.5 gal. (25 L) Buckets; optional additional Bucket |