missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Contec™ EasySat™ Bucketless Floor Mop

Eliminate the need for a bucket or wringer in cleanroom mopping. The Contec™ EasySat™ Bucketless Floor Mop features laser-cut sealed-edge VertiKlean™ mop head material and is fully autoclavable. A push button flow control releases cleaning solution on demand.

Supplier:  Contec™ BLMOP

Catalog No. 19-150-0692

Explore available promotions

Description

Description

  • Completely eliminates the need for bucket or wringer
  • Designed specifically for cleanroom use
  • Features lasercut sealed-edge VertiKlean™ mop head material, which is a lightweight polyester fabric laminated to clean ester foam for excellent abrasion resistance
  • System includes a 1.5L (52 oz.) translucent solution tank that can clean up to 5,000 square feet
  • Tank weighs only 3.2kg (7 lbs.) when full
  • Push button flow control releases cleaning solution on demand
  • Angled mop heads clean hard-to-reach areas and can be attached and removed within seconds
  • System and mop heads are fully autoclavable up to 121°C (250°F) for 30 minutes
  • Mop heads come in regular and textured styles and are also available gamma irradiated
  • Heads easily attaches to frame using traditional hook and loop fasteners

Compliance

AAMI ISO 11137:2006

Specifications

Specifications

Bucketless Floor Mop
Non-sterile
1.5 L Translucent Solution Tank that can clean up to 5,000 ft.2 Tank weighs only 3.2 kg when full
AAMI ISO 11137
21.5 in.
Autoclavable
Ideal for use with disinfectants, designed specifically for cleanroom floors
Polypropylene Copolymer, Silicone Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions