Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
Supplier:  Contec™ PS911EBETOH
Catalog No. 19-037-876
Explore available promotions
Description
Description
  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water or 70/30 blend of denaturated ethyl alcohol and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
Specifications
Specifications
Pre-saturated Wipe
28 cm
23 cm
Not Autoclavable
Single-use
Ideal for routine cleaning of tools, instruments and other laboratory equipment; designed for sterile environments
1-ply
Sterile
11 in.
11 in.
Polypropylene
ISO Class 4
28 x 23 cm (11 x 9 in.)
30 Wipes/Bag
70% Denatured Ethyl Alcohol/30% DI Water