missing translation for 'onlineSavingsMsg'
Learn More

Contec™ QuickConnect™ Mop Head Frames

Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.

$207.78 - $321.44

Specifications

Product Type Mop Head Frame
Sterility Non-sterile
Autoclavable Autoclavable
For Use With (Application) Designed for Use in Pharmaceutical and Medical Device Manufacturing Facilities, or Other Similar Critical or Controlled Environments.
View More Specs

Products 6
Catalog Number Mfr. No. Material For Use With (Equipment) Length (English) Price Quantity  
Catalog Number Mfr. No. Material For Use With (Equipment) Length (English) Price Quantity  
19-161-061
View Documents
Contec™
2651LF
Stainless Steel Regular VertiKlean MAX Head Frame 7.5 in.
Each for $293.01
Only null left
 
296367399
View Documents
Contec™
2653SS
Stainless Steel (304) Large VertiKlean Mop and VertiKlean MAX Head Frame 14 in.
Each for $321.44
Only null left
 
19-168-531
View Documents
Contec™
2646F
Stainless Steel (304) All Styles of Edgeless Mop Heads 7 in.
Each for $207.78
Only null left
 
19-168-536
View Documents
Contec™
2652LF
Stainless Steel Medium VertiKlean, Medium or Large VertiKlean MAX 11.5 in.
Each for $207.78
Only null left
 
19-159-261
View Documents
Contec™
2652SS
Stainless Steel VertiKlean MAX Mopping Systems 11.5 in.
Each for $268.01
Only null left
 
19168584
View Documents
Contec™
2651KLF
Stainless Steel Regular VertiKlean MAX Head Frame 7 in.
Each for $318.85
Only null left
 
Description

Description

Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.

  • Offer maximum flexibility for cleaning in critical environments
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
  • Capped end and quick-connect feature for easy attachment
Specifications

Specifications

Mop Head Frame
Non-sterile
Autoclavable
Designed for Use in Pharmaceutical and Medical Device Manufacturing Facilities, or Other Similar Critical or Controlled Environments.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'