Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads
Use with VertiKlean™ Wall Washing System which is designed for cleaning vertical surfaces. VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
Supplier:  Contec™ MVKMOPIR
 View more versions of this product
Catalog No. 19-120-825
Explore available promotions
Description
Description
  • Lightweight, polyester, disposable
  • Ideal for applying disinfectants and wet cleaning vertical surfaces, ceilings, and walls in controlled environments; reaching awkward corners and ceilings with a complete range of motion
  • Absorbent foam carries solvents and disinfectants to the surface; polyester fabric offers excellent abrasion resistance and helps distribute the liquid uniformly on the surface being cleaned
  • Heads are laser cut to seal edges, further limiting potential contamination in the mopping process
  • Textured surface option is ideal for applications requiring additional scrubbing performance
  • Contain no cellulose; compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
Specifications
Specifications
White
Sterile
Medium
12 in. Medium Frame
No
Mop Head
Autoclavable
Reusable
For applying disinfectants and wet cleaning ceilings and walls in controlled environments
Knitted Polyester, Polyester Foam