FUJIFILM Wako Control Standard Endotoxin
A lyophilized product composed of a 500 ng vial of endotoxin, purified from E. coli UKT-B strain that serves as an accurate and dependable standard for endotoxin testing.
Supplier:  FUJIFILM Wako CSE40375006
Catalog No. 12-333-027
Explore available promotions
Description
Description
  • Endotoxin derived from E. coli UKT-B
  • Can be used to prepare controls and standard curves
  • Potency of CSE supplied to match each lot of LAL
  • Reconstituted CSE can be stored at 2 to 10 °C for one month
Specifications
Specifications
Reconstituted CSE can be stored at 2 to 10 °C for one month
Lyophilized
6 vials containing 500 ng of CSE
500 ng