ILC Dover Conversion Kit - XL CBRN to HD PAPR
Convert the CBRN PAPR System into an HD system in a few easy steps. The blower stays the same, but the hood and filter selection provide flexibility depending on the application.
Supplier: ILC Dover S300029
Description
- Comes with the S-4001 Half Hood OV/AG cartridge filters
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
PAPR Conversion Kit | |
Half Hood and Cartridge Filters | |
Sentinel XL |