Corning™ XT Starter Ice-free Coolers
Bench top cooling systems that provide sample cooling or freezing without ice, electricity, or batteries.
Supplier:  Corning™ 432014
 View more versions of this product
Catalog No. 07-210-014
Explore available promotions
Description
Description
An open-platform ice-free cooler that accommodates most CoolRack™ and CoolSink™ modules. Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0.5° to 4.0°C) up to four hours. 1°C to 8°C temperature indicator provides visual assurance of temperature performance. To extend the cooling duration, keep an additional XT Cooling Core in the freezer and rotate the Cores as needed.
Specifications
Specifications
CoolRack and CoolSink Modules
Purple
1/Cs.
XT holder and XT Cooling Core
16.2 x 12.1 x 6 cm
Ice-free Cooler
1/Pk.
Sample cooling or freezing without ice, electricity, or batteries
1
Videos
SDS
Documents
Documents
Accessories