Corning™ XT Starter Ice-free Coolers
Bench top cooling systems that provide sample cooling or freezing without ice, electricity, or batteries.
Supplier:  Corning™ 432015
 View more versions of this product
Catalog No. 07-210-015
Explore available promotions
Description
Description
  • Open-platform cooler that accommodates most Corning CoolRack™ and Corning CoolSink™ modules
  • Low profile and small footprint make it ideal for use in the hood
  • Keeps samples cold (0.5°C to 4.0°C) for up to four hours 
  • 1°C to 8°C temperature indicator provides visual assurance of proper temperature. 
  • System may also be used with dry ice to create a compact, portable, snap-freezing workstation for bacteria, viruses, or proteins
Specifications
Specifications
CoolRack and CoolSink Modules
Purple
XT Starter holder only
16.2 x 12.1 x 6.1 cm
Ice-free Cooler
Sample cooling or freezing without ice, electricity, or batteries
1