Corning™ CoolRack™ 96 Storage Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
$1,110.17 - $1,487.73
Specifications
Type Storage Tube Module
View More Specs
Products 2
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
07-210-056
View Documents
Corning™
432056
96 x 0.5 mL 2D Bar Code Tubes N/A
07-210-057
View Documents
Corning™
432057
96 x 1.4 mL 2D Bar Code Tubes N/A
Description
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • 2D Barcode storage tube modules
  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.
  • Specifications
    Specifications
    Storage Tube Module
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'