CoorsTek™ Porcelain Capsule-Form Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
$46.33 - $67.90
Description
- Suitable for ash determinations, organic combustions, ignitions
- Capsule form with straight sides, flat bottom
- Glazed inside and out except on rim
Specifications
Porcelain |