Copan Diagnostics ESwab™ MRSA Collection System
Sample collection pack for various microbiology analyses
Supplier: Copan Diagnostics CA493CE02
Description
- Sterile
- Single use
- Pink Polypropylene screw-cap
- Applicator stick securely attaches to the cap after collection
- Tube with internal conical shape filled with 1mL of Liquid Amies Medium
- Sampling sites: Nose, throat, perineum
- 50 units per shelf pack
- 10 x 50 kits per box
Specifications
Sterile | |
Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 1 pink regular size flock swab plus 1 white regular size flocked swab | |
For various microbiology analysis | |
MRSA Collection System |