missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper foil, 0.254mm (0.010in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 91g/20 x 20cm

$145.32 - $447.97

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13379CU
View Documents
Thermo Scientific Chemicals
013379CU
20 x 20 cm
Each for $145.32
 
AA13379CV
View Documents
Thermo Scientific Chemicals
013379CV
20 x 100 cm
Each for $447.97
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

7440-50-8
8.94g/cm3
Cu
20 x 20 cm
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Odorless
Copper
Copper colored foil
99.9% (Metals basis)
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
Foil
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only