missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 000097GD

 View more versions of this product

Catalog No. AA00097GD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper foil
7440-50-8
Cu
50 x 500 mm
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
≥99.999%
Odorless
Foil
0.25mm (0.01 in.) thick, Hard, Temper: as rolled
(metals basis)
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
Puratronic™
≈11.2g/50x100mm

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only