missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper gauze, 20 mesh woven from 0.41mm (0.016in) dia wire, Thermo Scientific Chemicals

Wire Cloth

$54.67 - $223.41

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42777GT
View Documents
Thermo Scientific Chemicals
042777GT
150 x 150 mm
Each for $54.67
 
AA42777HB
View Documents
Thermo Scientific Chemicals
042777HB
300 x 300 mm
Each for $86.34
 
AA42777HZ
View Documents
Thermo Scientific Chemicals
042777HZ
300 x 1200 mm
Each for $223.41
 
Description

Description

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

7440-50-8
8.94g/cm3
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
Odorless
Copper
Copper colored guaze
Cu
150 x 150 mm
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
20 Mesh
Wire Cloth
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only