missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) acetate, 95%, Thermo Scientific Chemicals

$64.17 - $64.17

Chemical Identifiers

CAS 598-54-9
Molecular Formula C2H3CuO2
Molecular Weight (g/mol) 122.59
MDL Number MFCD00058908
InChI Key RFKZUAOAYVHBOY-UHFFFAOYSA-M
Synonym copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
PubChem CID 69023
IUPAC Name copper(1+);acetate
SMILES CC(=O)[O-].[Cu+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378810010
SDS View Documents
Thermo Scientific Chemicals
378810010
1 g Glass bottle
Each for $64.17
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

598-54-9
122.59
RFKZUAOAYVHBOY-UHFFFAOYSA-M
69023
CC(=O)[O-].[Cu+]
C2H3CuO2
MFCD00058908
copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
copper(1+);acetate

Specifications

250.0°C
White to Yellow
1 g
C2H3CuO2
MFCD00058908
copper i acetate, cuprous acetate, copper 1+ acetate, acetic acid, copper 1+ salt, unii-9u3n93lmzb, copper acetate, 9u3n93lmzb, acetic acid, copper 1+ salt 1:1, copper 1 acetate
RFKZUAOAYVHBOY-UHFFFAOYSA-M
copper(1+);acetate
69023
95%
Glass bottle
598-54-9
Powder
48.7% min. (Cu)
CH3C(O)OCu
15, 2648
Solubility in water: reacts
CC(=O)[O-].[Cu+]
122.59
122.59
Authentic
Copper(I) acetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-938-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only