missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
$277.87 - $277.87
Chemical Identifiers
CAS | 54678-23-8 |
---|---|
Molecular Formula | C2H6BrCuS |
Molecular Weight (g/mol) | 205.58 |
MDL Number | MFCD00043295 |
InChI Key | PMHQVHHXPFUNSP-UHFFFAOYSA-M |
Synonym | copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i |
PubChem CID | 2724234 |
SMILES | [Cu]Br.CSC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC213180250
|
Thermo Scientific Chemicals
213180250 |
25 g | Glass bottle |
Each for $277.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54678-23-8 | |
205.58 | |
PMHQVHHXPFUNSP-UHFFFAOYSA-M | |
2724234 |
C2H6BrCuS | |
MFCD00043295 | |
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i | |
[Cu]Br.CSC |
Specifications
54678-23-8 | |
Beige-Gray to White or Green | |
98.5% min. (Iodometry) | |
C2H6BrCuS | |
MFCD00043295 | |
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i | |
[Cu]Br.CSC | |
205.58 | |
205.58 | |
Crystalline Powder and/or Chunks |
132°C | |
Authentic | |
Glass bottle | |
(CH3)2S·CuBr | |
25 g | |
PMHQVHHXPFUNSP-UHFFFAOYSA-M | |
(methylsulfanyl)methane; bromocopper | |
2724234 | |
99% | |
Copper(I) bromide-dimethyl sulfide complex, 99% |
Safety and Handling
AIR SENSITIVE
MOISTURE SENSITIVE
EINECSNumber : 259-292-5
RUO – Research Use Only