Learn More
Copper(I) cyanide, 99%, extra pure, Thermo Scientific Chemicals
$150.33 - $366.22
Chemical Identifiers
CAS | 544-92-3 |
---|---|
Molecular Formula | CCuN |
Molecular Weight (g/mol) | 89.56 |
MDL Number | MFCD00010975 |
InChI Key | DOBRDRYODQBAMW-UHFFFAOYSA-N |
Synonym | copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper |
PubChem CID | 11009 |
IUPAC Name | copper(1+);cyanide |
SMILES | [C-]#N.[Cu+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC202082500
|
Thermo Scientific Chemicals
202082500 |
250 g | Plastic bottle |
Each for $150.33
|
|
||||
AC202080010
|
Thermo Scientific Chemicals
202080010 |
1 kg | Plastic bottle |
Each for $366.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
544-92-3 | |
89.56 | |
DOBRDRYODQBAMW-UHFFFAOYSA-N | |
11009 | |
[C-]#N.[Cu+] |
CCuN | |
MFCD00010975 | |
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper | |
copper(1+);cyanide |
Specifications
474.0°C | |
Beige-White to Green | |
250 g | |
CCuN | |
MFCD00010975 | |
15, 2651 | |
Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies, practically insoluble in alcohol | |
[C-]#N.[Cu+] | |
89.56 | |
89.56 | |
Extra Pure | |
2.92 | |
Copper(I) cyanide |
544-92-3 | |
Powder | |
98.5% min. (ex Cu) | |
CuCN | |
05,166; 06,146; 09,124; 16,97 | |
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper | |
DOBRDRYODQBAMW-UHFFFAOYSA-N | |
copper(1+);cyanide | |
11009 | |
99% | |
Plastic bottle | |
2.9200g/mL |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Rinse mouth.
GHS Signal Word: Danger
EINECSNumber : 208-883-6
RUO – Research Use Only