missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) iodide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 194900050

Catalog No. AC194900050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

C-X bond formation reagent
Specifications

Specifications

Copper(I) iodide
5kg
CuI
MFCD00010978
15, 2652
Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
190.45
Plastic Drum
7681-65-4
97.5% min. (Complexometry)
CuI
01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
190.45
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 231-674-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only