Learn More
Copper(I) trifluoromethanesulfonate benzene complex, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 371090010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Copper(I) trifluoromethanesulfonate benzene complex | |
7°C | |
90% | |
C8H6Cu2F6O6S2 | |
MFCD00043300 | |
12,144; 16,99 | |
Solubility in water: .. Other solubilities: soluble in benzene,toluene | |
503.33 |
42152-46-5 | |
Authentic | |
Glass bottle | |
(CF3SO3Cu)2C6H6 | |
1g | |
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex | |
503.33 | |
90% |
Chemical Identifiers
42152-46-5 | |
503.33 | |
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex |
C8H6Cu2F6O6S2 | |
MFCD00043300 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protecti
GHS Signal Word: Warning
EINECSNumber : 255-686-6
RUO â Research Use Only