missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1492014

 View more versions of this product

Catalog No. AAA1492014


Explore available promotions

Description

Description

Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Copper(II) 2,4-pentanedionate
∼286°C (decomposition)
Weak
MFCD00000016
4157957
Insoluble in water.
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
261.76
98%
13395-16-9
160°C (10 mmHg)
C10H14CuO4
25 g
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
261.76

Chemical Identifiers

13395-16-9
261.76
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
C10H14CuO4
MFCD00000016
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H360-H500

EINECSNumber : 236-477-9

RTECSNumber : GL6520000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only