Learn More
Copper(II) 2,4-pentanedionate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1492036
Description
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCopper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Copper(II) 2,4-pentanedionate | |
∼286°C (decomposition) | |
Weak | |
MFCD00000016 | |
4157957 | |
Insoluble in water. | |
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
261.76 | |
98% |
13395-16-9 | |
160°C (10 mmHg) | |
C10H14CuO4 | |
500 g | |
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II) | |
QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
261.76 |
Chemical Identifiers
13395-16-9 | |
261.76 | |
QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
C10H14CuO4 | |
MFCD00000016 | |
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II) | |
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H360-H500
EINECSNumber : 236-477-9
RTECSNumber : GL6520000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only