missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) Acetate Monohydrate 95.0+%, TCI America™

$59.25 - $144.17

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper(2+) diacetate hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C234625G
View Documents
TCI America
C234625G
25 g
Each for $59.25
 
C2346500G
View Documents
TCI America
C2346500G
500 g
Each for $144.17
 
Description

Description

Specifications

Chemical Identifiers

6046-93-1
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
165397
O.[Cu++].CC([O-])=O.CC([O-])=O
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
copper(2+) diacetate hydrate

Specifications

6046-93-1
Blue
C4H8CuO5
25 g
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
199.65
181.63
Crystalline Powder
115°C
5.2 to 5.5 (20g/L 20°C)
MFCD00149570
3077
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper(2+) diacetate hydrate
165397
≥95.0% (T)
Copper(II) Acetate Monohydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0693

RTECSNumber : AG3500000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'