Learn More
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110650250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) acetylacetonate | |
245.0°C | |
160.0°C (9.8 mmHg) | |
98% | |
C10H14CuO4 | |
25 g | |
2, 4-Pentanedione, metal derivative, Cupric acetylacetonate | |
Solubility in water: 0.2g/L (20°C). Other solubilities: 0.09g/100g alcohol (20°C),0.07g/100g benzene (20°C),readily soluble in chloroform,soluble in acetylacetone,dcm,dmf | |
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
261.76 | |
98% |
13395-16-9 | |
Blue to Blue-Gray | |
Authentic | |
Glass bottle | |
MFCD00000016 | |
02,81; 03,62 | |
160.0 °C | |
QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
261.76 | |
Crystalline Powder |
Chemical Identifiers
13395-16-9 | |
261.76 | |
QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
C10H14CuO4 | |
MFCD00000016 | |
2, 4-Pentanedione, metal derivative, Cupric acetylacetonate | |
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 236-477-9
RUO – Research Use Only