missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(II) Bis(2-hydroxyethyl)dithiocarbamate 98.0+%, TCI America™
$98.75 - $296.25
Chemical Identifiers
CAS | 52611-57-1 |
---|---|
Molecular Formula | C10H20CuN2O4S4 |
Molecular Weight (g/mol) | 424.066 |
MDL Number | MFCD00059149 |
InChI Key | CWZKOKSQKMDNMW-UHFFFAOYSA-L |
Synonym | Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt |
PubChem CID | 504564 |
IUPAC Name | copper;N,N-bis(2-hydroxyethyl)carbamodithioate |
SMILES | C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Cu+2] |
Chemical Identifiers
52611-57-1 | |
424.066 | |
CWZKOKSQKMDNMW-UHFFFAOYSA-L | |
504564 | |
C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Cu+2] |
C10H20CuN2O4S4 | |
MFCD00059149 | |
Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt | |
copper;N,N-bis(2-hydroxyethyl)carbamodithioate |
Specifications
52611-57-1 | |
C10H20CuN2O4S4 | |
5 g | |
CWZKOKSQKMDNMW-UHFFFAOYSA-L | |
copper;N,N-bis(2-hydroxyethyl)carbamodithioate | |
504564 | |
≥98.0% (T) | |
Copper(II) Bis(2-hydroxyethyl)dithiocarbamate |
Black-Green | |
MFCD00059149 | |
Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt | |
C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Cu+2] | |
424.066 | |
424.07 | |
Crystalline Powder |
Safety and Handling
TSCA : No