missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206330050

 View more versions of this product

Catalog No. AC206330050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

900.0°C
498.0°C
Black-Gray to Gray
5 g
Br2Cu
MFCD00010970
15, 2619
Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4
223.36
Extra Pure
4.77
Copper(II) bromide
7789-45-9
Crystalline Powder
99% min. (Iodometry)
CuBr2
01,161; 02,84; 05,158; 06,138; 13,112; 15,100
Cupric bromide
223.36
≥99%
Glass bottle

Chemical Identifiers

7789-45-9
223.36
Cupric bromide
Br2Cu
MFCD00010970
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 232-167-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only