Learn More
Copper(II) bromide, 99%
CAS: 7789-45-9 | CuBr2
$65.67 - $333.64
Chemical Identifiers
| CAS | 7789-45-9 |
|---|---|
| Molecular Formula | CuBr2 |
| MDL Number | MFCD00010970 |
| Synonym | Cupric bromide |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1633514
|
Thermo Scientific Chemicals
A1633514 |
25 g |
Each for $65.67
|
|
|||||
|
AAA1633522
|
Thermo Scientific Chemicals
A1633522 |
100 g |
Each for $107.52
|
|
|||||
|
AAA1633536
|
Thermo Scientific Chemicals
A1633536 |
500 g |
Each for $333.64
|
|
|||||
Description
Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCopper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.
Solubility
Soluble in water, alcohol, ammonia and acetone. Insoluble in benzene, ether, sulfuric acid and ethyl ether.
Notes
Hygroscopic. Incompatible with oxidizing agents and metals.
Chemical Identifiers
| 7789-45-9 | |
| CuBr2 | |
| MFCD00010970 | |
| Cupric bromide |
Specifications
| 900°C | |
| 7789-45-9 | |
| CuBr2 | |
| UN3260 | |
| Cupric bromide | |
| 223.36 | |
| Odorless | |
| 4.71 |
| 498°C | |
| 25 g | |
| MFCD00010970 | |
| 14,2629 | |
| Soluble in water,alcohol,ammonia and acetone. Insoluble in benzene,ether,sulfuric acid and ethyl ether. | |
| 99% | |
| Hygroscopic | |
| Copper(II) bromide |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-167-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only