Learn More
Copper(II) carbonate dihydroxide, Cu 55% min, Thermo Scientific Chemicals
$138.42 - $388.97
Chemical Identifiers
CAS | 12069-69-1 |
---|---|
Molecular Formula | CH2Cu2O5 |
MDL Number | MFCD00010976 |
Synonym | Copper(II) carbonate, basic, Copper(II) hydroxide carbonate |
Description
Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12069-69-1 | |
CH2Cu2O5 | |
MFCD00010976 | |
Copper(II) carbonate, basic, Copper(II) hydroxide carbonate |
Specifications
200°C (decomposition) | |
12069-69-1 | |
500 g | |
MFCD00010976 | |
Soluble in acid,ammonia,ammonium salt and alkali metal carbonate solution. Insoluble in water and alcohol. | |
4 |
Green | |
Powder | |
CH2Cu2O5 | |
Copper(II) carbonate, basic, Copper(II) hydroxide carbonate | |
221.11 | |
Copper(II) carbonate dihydroxide, Cu 55% min. |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302+H332-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 235-113-6
RTECSNumber : GL6910000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only