missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) chloride dihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1606430

 View more versions of this product

Catalog No. AAA1606430


Explore available promotions

Description

Description

Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.

Solubility
Soluble in water, methanol and ethanol.

Notes
Incompatible with materials such as oxidizing agents, metals and acids.
Specifications

Specifications

Copper(II) chloride dihydrate
10125-13-0
250 g
MFCD00149674
14,2633
Soluble in water,methanol and ethanol.
O.O.[Cl-].[Cl-].[Cu++]
170.48
CHEBI:86318
99%
Hygroscopic
100°C (decomposition)
3.0 to 3.8
Cl2CuH4O2
UN2802
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
dichlorocopper;dihydrate
61482
170.48 (134.45 Anhydrous)
Odorless
2.51

Chemical Identifiers

10125-13-0
170.48
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
61482
dichlorocopper;dihydrate
Cl2CuH4O2
MFCD00149674
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
CHEBI:86318
O.O.[Cl-].[Cl-].[Cu++]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: COPPER CHLORIDE

EINECSNumber : 231-210-2

RTECSNumber : GL7030000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only