Learn More
Copper(II) formate tetrahydrate, 98%, Thermo Scientific Chemicals
$86.44 - $300.90
Chemical Identifiers
CAS | 5893-61-8 |
---|---|
Molecular Formula | C2H10CuO8 |
Molecular Weight (g/mol) | 225.64 |
MDL Number | MFCD00167128 |
InChI Key | LSIWWRSSSOYIMS-UHFFFAOYSA-L |
Synonym | copper ii formate tetrahydrate, cupric formate tetrahydrate, copper diformate tetrahydrate, unii-v7ra28204j, copper 2+ formate tetrahydrate, cupric formate tetrahydrate mi, copper formate cu hcoo 2 tetrahydrate, formic acid copper salt 2:1 tetrahydrate, acmc-209m8c |
PubChem CID | 6432244 |
IUPAC Name | copper;diformate;tetrahydrate |
SMILES | C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1856918
|
Thermo Scientific Chemicals
A1856918 |
50 g |
Each for $86.44
|
|
|||||
AAA1856930
|
Thermo Scientific Chemicals
A1856930 |
250 g |
Each for $300.90
|
|
|||||
Description
It is employed as an antibacterial agent in the treatment of cellulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an antibacterial agent in the treatment of cellulose.
Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
5893-61-8 | |
225.64 | |
LSIWWRSSSOYIMS-UHFFFAOYSA-L | |
6432244 | |
C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2] |
C2H10CuO8 | |
MFCD00167128 | |
copper ii formate tetrahydrate, cupric formate tetrahydrate, copper diformate tetrahydrate, unii-v7ra28204j, copper 2+ formate tetrahydrate, cupric formate tetrahydrate mi, copper formate cu hcoo 2 tetrahydrate, formic acid copper salt 2:1 tetrahydrate, acmc-209m8c | |
copper;diformate;tetrahydrate |
Specifications
5893-61-8 | |
MFCD00167128 | |
UN3077 | |
copper ii formate tetrahydrate, cupric formate tetrahydrate, copper diformate tetrahydrate, unii-v7ra28204j, copper 2+ formate tetrahydrate, cupric formate tetrahydrate mi, copper formate cu hcoo 2 tetrahydrate, formic acid copper salt 2:1 tetrahydrate, acmc-209m8c | |
LSIWWRSSSOYIMS-UHFFFAOYSA-L | |
copper;diformate;tetrahydrate | |
6432244 | |
98% |
C2H10CuO8 | |
50 g | |
14,2639 | |
Soluble in water (125g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents. | |
C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2] | |
225.64 | |
225.65 (153.59 Anhydrous) | |
Copper(II) formate tetrahydrate |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 208-865-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only