missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) gluconate, Thermo Scientific Chemicals

$59.18 - $580.73

Chemical Identifiers

CAS 527-09-3
Molecular Formula C12H22CuO14
Molecular Weight (g/mol) 453.84
MDL Number MFCD00075297
InChI Key OCUCCJIRFHNWBP-IYEMJOQQSA-L
Synonym copper gluconate
PubChem CID 131854750
ChEBI CHEBI:31431
IUPAC Name copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1532518
View Documents
Thermo Scientific Chemicals
01532518
50 g
Each for $59.18
 
AA1532530
View Documents
Thermo Scientific Chemicals
01532530
250 g
Each for $193.58
 
AA15325A1
View Documents
Thermo Scientific Chemicals
015325A1
1 kg
Each for $580.73
 
Description

Description

Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

527-09-3
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
131854750
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C12H22CuO14
MFCD00075297
copper gluconate
CHEBI:31431
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]

Specifications

527-09-3
C12H22CuO14
50 g
copper gluconate
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
131854750
453.85
Copper(II) gluconate
155°C to 157°C
MFCD00075297
14,2640
Soluble in water. Insoluble in acetone,alcohol and ether.
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]
453.84
CHEBI:31431
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-408-2

RTECSNumber : LZ5058000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only