Learn More
Copper(II) hydroxide, tech. 94%, stab., Thermo Scientific Chemicals
Used As a Raw Material in Chemical Processing and Catalyst Manufacturing
Supplier: Thermo Scientific Chemicals 03273336
Description
Used as a raw material in chemical processing and catalyst manufacturing. Copper(II) hydroxide is used as ceramic colorant. In organic synthesis, it is used as a catalyst to prepare 1-((2-aminoethyl)amino)anthraquinone, 1-amino-4-((2-aminoethyl)amino)anthraquinone by reacting with 1-bromoanthraquinone or 1-amino-4-bromoanthraquinone respectively. It is involved in the synthesis of benzoic acid and octanoic acid. It is used as a precursor in the production of rayon, battery electrodes and other copper salts It is also used as a dye mordant, paper colorant, pigment and feed additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) hydroxide | |
20427-59-2 | |
500 g | |
H2CuO2 | |
Slightly soluble in waterSoluble in acids,ammonium hydroxide,dilute hydrochloric acid,concentrated alkali and potassium cyanide. Insoluble in water,ethanol,and acetone. | |
Technical | |
Hygroscopic | |
3.4 g/cm3 |
Blue | |
Powder | |
94% | |
MFCD00010968 | |
97.55 | |
Odorless | |
Blue powder |
Chemical Identifiers
20427-59-2 | |
H2CuO2 | |
MFCD00010968 |
Safety and Handling
P260-P264b-P270-P271-P280-P284-P301+P312-P304+P340-P305+P351+P338-P310-P330-P501c
H302-H318-H330
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 243-815-9
RTECSNumber : GL7600000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only