Learn More
Copper(II) methoxide, typically 98% (metals basis), Thermo Scientific Chemicals
$56.45 - $746.55
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
AA1886103
|
Thermo Scientific Chemicals
01886103 |
1 g |
chaque for $56.45
|
|
|||||
AA1886106
|
Thermo Scientific Chemicals
01886106 |
5 g |
chaque for $184.96
|
|
|||||
AA1886114
|
Thermo Scientific Chemicals
01886114 |
25 g |
chaque for $746.55
|
|
|||||
Description
Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
1184-54-9 | |
MFCD00015611 |
Spécifications
1184-54-9 | |
Green | |
Cu(OCH3)2 | |
1 g | |
Moisture sensitive | |
125.61 | |
Powder | |
(metals basis) |
∼206°C (decomposition) | |
Alcohol-like | |
MFCD00015611 | |
UN3181 | |
Reacts with water. | |
98% (typically) | |
Copper(II) methoxide |
Sécurité et manipulation
GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only