Learn More
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
$61.73 - $1568.68
Chemical Identifiers
CAS | 13778-31-9 |
---|---|
Molecular Formula | CuN2O6 |
Molecular Weight (g/mol) | 187.55 |
MDL Number | MFCD00149669 |
InChI Key | XTVVROIMIGLXTD-UHFFFAOYSA-N |
Synonym | copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci |
PubChem CID | 15303652 |
SMILES | [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA1069906
|
Thermo Scientific Chemicals
01069906 |
5 g |
Each for $61.73
|
|
||||||
AA1069914
|
Thermo Scientific Chemicals
01069914 |
25 g |
Each for $140.25
|
|
||||||
AA1069922
|
Thermo Scientific Chemicals
01069922 |
100 g |
Each for $440.11
|
|
||||||
AA1069936
|
Thermo Scientific Chemicals
01069936 |
500 g |
Each for $1,568.68
|
|
||||||
Description
It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13778-31-9 | |
187.55 | |
XTVVROIMIGLXTD-UHFFFAOYSA-N | |
15303652 |
CuN2O6 | |
MFCD00149669 | |
copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci | |
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
13778-31-9 | |
Crystalline Aggregates | |
99.999% (Metals basis) | |
MFCD00149669 | |
Soluble in ethanol and water. Insoluble in ethyl acetate. | |
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
187.55 | |
187.55 (Anhydrous) | |
Hygroscopic |
Blue | |
5 g | |
CuN2O6 | |
copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci | |
XTVVROIMIGLXTD-UHFFFAOYSA-N | |
copper(2+) dinitrate | |
15303652 | |
Odorless | |
Copper(II) nitrate hydrate |
Safety and Handling
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 221-838-5
RTECSNumber : GL7875000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only