missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) oxalate hemihydrate, 98%, Thermo Scientific Chemicals

$115.30 - $364.38

Chemical Identifiers

CAS 5893-66-3
Molecular Formula C4H8Cu2O10
Molecular Weight (g/mol) 343.19
MDL Number MFCD00150451
InChI Key NNZNHNMQOYUANB-UHFFFAOYSA-N
Synonym cupric oxalate, copper oxalate hemihydrate
PubChem CID 122130000
IUPAC Name copper;oxalic acid;hydrate
SMILES O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1536518
View Documents
Thermo Scientific Chemicals
A1536518
50 g
Each for $115.30
 
AAA1536530
View Documents
Thermo Scientific Chemicals
A1536530
250 g
Each for $364.38
 
Description

Description

Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
Specifications

Chemical Identifiers

5893-66-3
343.19
NNZNHNMQOYUANB-UHFFFAOYSA-N
122130000
O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O
C4H8Cu2O10
MFCD00150451
cupric oxalate, copper oxalate hemihydrate
copper;oxalic acid;hydrate

Specifications

5893-66-3
C4H8Cu2O10
50 g
14,2645
Soluble in NH4OH
O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O
343.19
160.57 (151.51 Anhydrous)
Powder
98%
MFCD00150451
UN3288
cupric oxalate, copper oxalate hemihydrate
NNZNHNMQOYUANB-UHFFFAOYSA-N
copper;oxalic acid;hydrate
122130000
98%
Copper(II) Oxalate Hemihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 212-411-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only