missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) phosphate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1736036

 View more versions of this product

Catalog No. AAA1736036


Explore available promotions

Description

Description

Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
Specifications

Specifications

Copper(II) phosphate
7798-23-4
Cu3O8P2
MFCD00036268
copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
GQDHEYWVLBJKBA-UHFFFAOYSA-H
tricopper;diphosphate
86469
98%
>300°C
500 g
Cu3(PO4)2
14,2648
Insoluble with water and ethanol. Soluble in ammonia,ammonium hydroxide,acetone.
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]
380.578
380.58
Odorless

Chemical Identifiers

7798-23-4
380.578
GQDHEYWVLBJKBA-UHFFFAOYSA-H
86469
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]
Cu3O8P2
MFCD00036268
copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
tricopper;diphosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 232-254-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only