missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) phthalocyanine, Thermo Scientific Chemicals

$63.89 - $703.78

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160; Pigment Blue 15
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4365009
View Documents
Thermo Scientific Chemicals
04365009
10 g
Each for $63.89
 
AA4365018
View Documents
Thermo Scientific Chemicals
04365018
50 g
Each for $213.30
 
AA4365030
View Documents
Thermo Scientific Chemicals
04365030
250 g
Each for $703.78
 
Description

Description

Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

147-14-8
C32H16CuN8
MFCD00010719
C.I. 74160; Pigment Blue 15

Specifications

147-14-8
Blue to Purple
MFCD00010719
4121848
C.I. 74160; Pigment Blue 15
576.08
Copper (II) phthalocyanine
350°C
C32H16CuN8
10 g
14,2520
It is insoluble in water.
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-685-1

RTECSNumber : GL8510000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only