Learn More
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%
CAS: 16570-28-8 | Cu2O7P2 | 301.03 g/mol
$148.12 - $559.60
Chemical Identifiers
| CAS | 16570-28-8 |
|---|---|
| Molecular Formula | Cu2O7P2 |
| Molecular Weight (g/mol) | 301.03 |
| MDL Number | MFCD00150225 |
| InChI Key | PEVJCYPAFCUXEZ-UHFFFAOYSA-J |
| IUPAC Name | dicopper(2+) (phosphonatooxy)phosphonate |
| SMILES | [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O |
Description
Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16570-28-8 | |
| 301.03 | |
| PEVJCYPAFCUXEZ-UHFFFAOYSA-J | |
| [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O |
| Cu2O7P2 | |
| MFCD00150225 | |
| dicopper(2+) (phosphonatooxy)phosphonate |
Specifications
| Blue | |
| Powder | |
| Cu2O7P2 | |
| Insoluble in water. | |
| [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O | |
| 301.03 | |
| P2O7 43.5 to 48.5% |
| 16570-28-8 | |
| 1 kg | |
| MFCD00150225 | |
| PEVJCYPAFCUXEZ-UHFFFAOYSA-J | |
| dicopper(2+) (phosphonatooxy)phosphonate | |
| 301.02 (Anhydrous) | |
| Copper (II) pyrophosphate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-250-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only