missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) sulfate, 98%, pure, anhydrous, Thermo Scientific Chemicals

$88.34 - $310.41

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.6
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC422870050
SDS View Documents
Thermo Scientific Chemicals
422870050
5 g Glass bottle
Each for $88.34
 
AC422871000
SDS View Documents
Thermo Scientific Chemicals
422871000
100 g Glass bottle
Each for $100.52
 
AC422875000
SDS View Documents
Thermo Scientific Chemicals
422875000
500 g Glass bottle
Each for $310.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-98-7
159.6
ARUVKPQLZAKDPS-UHFFFAOYSA-L
24462
copper;sulfate
CuO4S
MFCD00010981
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
CHEBI:23414
[O-]S(=O)(=O)[O-].[Cu+2]

Specifications

200.0°C
Gray
5 g
CuO4S
MFCD00010981
15,2643
Solubility in water: 203g/L (20°C).
[O-]S(=O)(=O)[O-].[Cu+2]
159.6
CHEBI:23414
98%
Glass bottle
7758-98-7
Powder
97.5% min. (Iodometry)
CuSO4
01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper;sulfate
24462
159.6
Pure
Copper(II) sulfate, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y

GHS Signal Word: Warning

EINECSNumber : 231-847-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only